Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gm14483 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001111037.1 | NP_001104507.1 | 1 | 470 | ||
GenBank mRNA | BC087913.1 | - | 1 | 470 | 6705184 |
Gene Symbol: 1700012L04Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029588.3 | NP_083864.1 | 1 | 501 | ||
GenBank mRNA | AK005922.1 | - | 1 | 503 |
Gene Symbol: Gm14474 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242947.1 | NP_001229876.1 | 1 | 463 |
Gene Symbol: Gm14482 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242952.1 | NP_001229881.1 | 1 | 475 |
Gene Symbol: Gm14484 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025260.2 | NP_001020431.1 | 1 | 463 | ||
GenBank mRNA | BC049545.1 | - | 463 | 6770892 |
Gene Symbol: Gm14501 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001085537.2 | NP_001079006.1 | 1 | 503 |