Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spice1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144550.4 | NP_653133.3 | 9, 10 | 996 | ||
GenBank mRNA | AK048789.1 | - | 9, 10 | 940 | ||
AY099108.1 | - | 9, 10 | 1276 | |||
BC024956.1 | - | 1, 2 | 350 | |||
BC052381.1 | - | 9, 10 | 950 | 5703283 | ||
BC056960.1 | - | 9, 10 | 950 | 5703283 |