Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gast | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010257.3 | NP_034387.3 | 3 | 361 | ||
GenBank mRNA | AK008062.1 | - | 3 | 362 | ||
AK008159.1 | - | 3 | 362 | |||
AK008313.1 | - | 3 | 362 | |||
AK008420.1 | - | 3 | 362 | |||
AK008494.1 | - | 3 | 362 | |||
AK008649.1 | - | 3 | 365 | |||
BC064791.1 | - | 3 | 362 | 6431366 | ||
X94758.1 | - | 3 | 366 |