Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Xlr4b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001293676.1 | NP_001280605.1 | 9 | 945 | ||
NM_001293677.1 | NP_001280606.1 | 9 | 898 | |||
NM_001293678.1 | NP_001280607.1 | 9 | 922 | |||
NM_001293679.1 | NP_001280608.1 | 9 | 934 | |||
NM_001293680.1 | NP_001280609.1 | 9 | 958 | |||
NM_021365.3 | NP_067340.2 | 9 | 1062 | |||
GenBank mRNA | AL136330.1 | - | 9 | 1029 | ||
BC025576.1 | - | 9 | 898 | 5323621 | ||
BC034848.1 | - | 9 | 1486 | |||
BC138975.1 | - | 9 | 804 | 8861997 | ||
BC138979.1 | - | 9 | 828 | 8862001 | ||
BC145481.1 | - | 9 | 828 | 9054044 | ||
BC145482.1 | - | 9 | 840 | 9054045 | ||
BC145483.1 | - | 9 | 864 | 9054046 |