Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sin3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113248.2 | NP_001106719.1 | 3 | 352 | ||
NM_009188.4 | NP_033214.2 | 3 | 352 | |||
GenBank mRNA | AF038848.1 | - | 3 | 332 | ||
AK146847.1 | - | 3 | 335 | |||
AK159076.1 | - | 3 | 335 | |||
AK165592.1 | - | 3 | 337 | |||
AK169433.1 | - | 3 | 337 | |||
AK173005.1 | - | 3 | 321 | |||
AK187217.1 | - | 2 | 186 | |||
BC020049.1 | - | 3 | 316 | |||
BC021160.1 | - | 3 | 335 | |||
BC096367.1 | - | 3 | 320 | |||
BC129859.1 | - | 3 | 324 | |||
L38622.2 | - | 3 | 324 |