Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CAMTA1 SIRNA Assays ›
Gene Symbol:
CAMTA1
Gene Name:
calmodulin binding transcription activator 1
Gene Aliases:
CANPMR
Chromosome Location:
Chr.1: 6785324 - 7769706 on Build GRCh38
UniGene ID:
Hs.397705
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s23420
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CAMTA1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001195563.1
NP_001182492.1
3
386
NM_001242701.1
NP_001229630.1
3
386
NM_015215.3
NP_056030.1
3
386
NR_038934.1
2
316
XM_011541083.2
XP_011539385.1
3
250
XM_011541084.2
XP_011539386.1
3
250
XM_011541085.2
XP_011539387.1
4
322
XM_011541086.2
XP_011539388.1
3
250
XM_011541087.2
XP_011539389.1
3
250
XM_011541088.2
XP_011539390.1
2
180
XM_011541089.2
XP_011539391.1
3
250
XM_011541090.2
XP_011539392.1
3
250
XM_011541091.2
XP_011539393.1
3
250
XM_011541092.2
XP_011539394.1
3
250
XM_017000773.1
XP_016856262.1
4
1002
XM_017000774.1
XP_016856263.1
3
250
XM_017000775.1
XP_016856264.1
2
180
XM_017000776.1
XP_016856265.1
3
239
XM_017000777.1
XP_016856266.1
3
250
XM_017000778.1
XP_016856267.1
3
250
XM_017000779.1
XP_016856268.1
2
180
XM_017000780.1
XP_016856269.1
3
250
XM_017000781.1
XP_016856270.1
3
250
GenBank mRNA
AB020640.3
-
3
386
AY037153.1
-
3
225
BC116457.1
-
3
232
BC151835.1
-
3
386
8860115
BG027385.1
-
3
247
BM470492.1
-
177
CD103791.1
-
3
185
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_001195559, NM_001195560, BF290447, BC106025.
This siRNA also targets the following transcripts in Mouse: NM_001081557, NM_001195565, AK158710, AK034038, AK009988, and others.
Panther Classification:
Molecular Function -
DNA-binding transcription factor
Gene Ontology Categories:
Function(s)
Process(es)
transcription from RNA polymerase II promoter
positive regulation of transcription from RNA polymerase II promoter
neuromuscular process controlling balance
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
sequence-specific DNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-65d5d77f49-g5d4h:80/100.66.76.71:80.
git-commit: a4c025c8ba47546866237b65de1f276a5fdbbe78
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.18.1-Offline