Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ifna11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008333.2 | NP_032359.2 | 1 | 595 |
Gene Symbol: Ifna1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010502.2 | NP_034632.2 | 1 | 555 |
Gene Symbol: Ifna16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_206867.1 | NP_996750.1 | 1 | 555 | ||
GenBank mRNA | BC114392.1 | - | 1 | 554 |