Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gm6182 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK008984.1 | - | 6 | 1000 | ||
AK166086.1 | - | 5 | 903 | |||
BC048160.1 | - | 5 | 866 | |||
BC051922.1 | - | 5 | 866 | |||
BC059210.1 | - | 4 | 2000 |
Gene Symbol: 0610010B08Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177543.1 | NP_001171014.1 | 1200 |
Gene Symbol: Gm14305 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099327.1 | NP_001092797.1 | 5 | 1108 |
Gene Symbol: Gm14308 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099349.2 | NP_001092819.1 | 5 | 1200 |
Gene Symbol: Gm14391 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099308.2 | NP_001092778.1 | 5 | 1264 |
Gene Symbol: Gm14430 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100415.1 | NP_001093885.1 | 1200 |
Gene Symbol: Gm14432 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100416.2 | NP_001093886.1 | 1108 |
Gene Symbol: Gm14434 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001101804.1 | NP_001095274.1 | 1200 |
Gene Symbol: Gm4724 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256480.1 | NP_001243409.1 | 1200 |
Gene Symbol: Gm6710 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164689.1 | NP_001158161.1 | 5 | 1257 |