Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gm14393 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001085546.2 | NP_001079015.1 | 4 | 350 | ||
GenBank mRNA | CF161713.1 | - | 4 | 350 | ||
CF164689.1 | - | 4 | 344 |
Gene Symbol: 0610010B08Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177543.1 | NP_001171014.1 | 415 |
Gene Symbol: Gm14305 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099327.1 | NP_001092797.1 | 5 | 407 |
Gene Symbol: Gm14308 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099349.2 | NP_001092819.1 | 5 | 415 |
Gene Symbol: Gm14326 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190302.2 | NP_001177231.1 | 5 | 416 | ||
GenBank mRNA | CV555523.1 | - | 3 | 186 |
Gene Symbol: Gm14391 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099308.2 | NP_001092778.1 | 5 | 479 |
Gene Symbol: Gm14430 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100415.1 | NP_001093885.1 | 415 |
Gene Symbol: Gm14432 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100416.2 | NP_001093886.1 | 407 | |||
GenBank mRNA | CV559949.1 | - | 407 |
Gene Symbol: Gm14434 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001101804.1 | NP_001095274.1 | 415 |
Gene Symbol: Gm4724 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256480.1 | NP_001243409.1 | 415 |
Gene Symbol: Gm6710 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164689.1 | NP_001158161.1 | 5 | 472 |