Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il11ra1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163401.1 | NP_001156873.1 | 5 | 542 | ||
NM_001172054.1 | NP_001165525.1 | 6 | 628 | |||
NM_010549.3 | NP_034679.1 | 5 | 547 | |||
GenBank mRNA | AK143811.1 | - | 3 | 1414 | ||
AK146742.1 | - | 5 | 495 | |||
AK155407.1 | - | 5 | 489 | |||
AK159559.1 | - | 5 | 477 | |||
BC004619.1 | - | 5 | 466 | 3585497 | ||
BC057664.1 | - | 5 | 477 | 3668288 | ||
BC069984.1 | - | 5 | 529 | 6508165 | ||
U14412.1 | - | 5 | 477 | |||
X74953.1 | - | 5 | 500 |
Gene Symbol: Gm13305 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099348.1 | NP_001092818.1 | 733 |
Gene Symbol: Gm2002 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100596.1 | NP_001094066.1 | 6 | 734 |
Gene Symbol: Il11ra2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010550.3 | NP_034680.3 | 6 | 734 | ||
GenBank mRNA | BC132549.1 | - | 5 | 533 | 40130826 | |
BC138592.1 | - | 5 | 533 | 8861612 | ||
BC145341.1 | - | 5 | 533 | 9053897 | ||
U69491.1 | - | 6 | 734 | |||
X98519.1 | - | 6 | 746 |