Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gm5797 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025085.2 | NP_001020256.1 | 5 | 860 | ||
GenBank mRNA | BC090991.1 | - | 5 | 860 | 6742820 |
Gene Symbol: 4930555G01Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175393.4 | NP_780602.2 | 5 | 1711 | ||
GenBank mRNA | AK076920.1 | - | 5 | 1712 |
Gene Symbol: Gm10413 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029288.3 | NP_083564.2 | 5 | 860 | ||
GenBank mRNA | AK005583.1 | - | 5 | 850 | ||
AV258319.1 | - | 1 | 149 | |||
BC100412.1 | - | 5 | 860 | 30916569 |
Gene Symbol: Gm5458 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024706.2 | NP_001019877.2 | 5 | 866 | ||
GenBank mRNA | BC060995.1 | - | 5 | 754 | 6774503 |
Gene Symbol: Gm5795 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270806.1 | NP_001257735.1 | 5 | 831 |