Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cd300lh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199201.1 | NP_954671.1 | 2, 3 | 380 | ||
GenBank mRNA | AY457049.1 | - | 2, 3 | 380 | ||
BC119352.1 | - | 2, 3 | 415 | 8734101 | ||
BC120623.1 | - | 415 | 40129546 |
Gene Symbol: Gm11710 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001101656.2 | NP_001095126.1 | 415 |
Gene Symbol: Gm11711 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001101657.2 | NP_001095127.1 | 2, 3 | 415 |