Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mndal | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170853.1 | NP_001164324.1 | 1250 | |||
GenBank mRNA | AK087931.1 | - | 6 | 1074 | ||
AK136391.1 | - | 1252 | ||||
AK161717.1 | - | 3 | 965 | |||
FJ594740.1 | - | 6 | 1149 | |||
FJ594741.1 | - | 1158 |
Gene Symbol: Ifi204 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008329.2 | NP_032355.2 | 5 | 995 | ||
GenBank mRNA | BC010546.1 | - | 973 | 4018506 | ||
CT010351.1 | - | 887 | ||||
M31419.1 | - | 1056 |
Gene Symbol: Ifi205 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172648.3 | NP_766236.2 | 5 | 955 | ||
GenBank mRNA | AK040191.1 | - | 5 | 885 | ||
BC040425.1 | - | 5 | 955 | 3466347 | ||
CT010211.1 | - | 4 | 803 | |||
M74124.1 | - | 4 | 736 |
Gene Symbol: Mnda | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033450.4 | NP_001028622.1 | 5 | 1119 | ||
NM_001301745.1 | NP_001288674.1 | 5 | 1113 | |||
GenBank mRNA | AK152278.1 | - | 5 | 987 | ||
AK152683.1 | - | 5 | 987 | |||
AK152786.1 | - | 5 | 996 | |||
AK166217.1 | - | 5 | 1119 | |||
BC132314.1 | - | 5 | 940 | 40130591 | ||
BC132316.1 | - | 5 | 940 | 40130593 | ||
BC145397.1 | - | 5 | 934 | 9053955 | ||
M74123.1 | - | 5 | 974 | |||
S62227.1 | - | 5 | 974 |