Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mnda | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033450.4 | NP_001028622.1 | 2 | 298 | ||
NM_001301745.1 | NP_001288674.1 | 2 | 298 | |||
GenBank mRNA | AK080485.1 | - | 2 | 247 | ||
AK152278.1 | - | 2 | 167 | |||
AK152683.1 | - | 2 | 166 | |||
AK152786.1 | - | 2 | 175 | |||
AK166217.1 | - | 2 | 298 | |||
BC132314.1 | - | 2 | 119 | 40130591 | ||
BC132316.1 | - | 2 | 119 | 40130593 | ||
BC145397.1 | - | 2 | 119 | 9053955 | ||
CJ067068.1 | - | 2 | 290 | |||
M74123.1 | - | 2 | 153 | |||
S62227.1 | - | 2 | 153 |
Gene Symbol: Ifi204 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008329.2 | NP_032355.2 | 2 | 174 | ||
GenBank mRNA | AK138785.1 | - | 2 | 199 | ||
AK150522.1 | - | 2 | 175 | |||
BC010546.1 | - | 131 | 4018506 | |||
CT010351.1 | - | 45 | ||||
M31419.1 | - | 172 |