Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZC3H7B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017590.5 | NP_060060.3 | 21 | 2672 | ||
XM_011530016.2 | XP_011528318.1 | 21 | 2594 | |||
GenBank mRNA | AB028954.1 | - | 18 | 2285 | ||
AK315009.1 | - | 21 | 2614 | |||
BC024313.2 | - | 12 | 1339 | |||
BC098378.1 | - | 21 | 2610 | |||
BC152558.1 | - | 21 | 2650 | 8860405 |