Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gm3285 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001101631.1 | NP_001095101.1 | 1 | 244 |
Gene Symbol: Gm10318 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001162944.1 | NP_001156416.1 | 1 | 233 |
Gene Symbol: Gm19402 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001205032.1 | NP_001191961.1 | 1 | 335 |
Gene Symbol: Gm2696 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001205009.1 | NP_001191938.1 | 1 | 260 |
Gene Symbol: Gm3238 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001101630.1 | NP_001095100.1 | 1 | 230 |
Gene Symbol: Krtap10-10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024709.3 | NP_001019880.2 | 1 | 297 | ||
GenBank mRNA | AK079449.1 | - | 1 | 298 |
Gene Symbol: Krtap10-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135991.1 | NP_001129463.1 | 1 | 303 | ||
GenBank mRNA | AK020700.1 | - | 1 | 330 | ||
BC115499.1 | - | 1 | 303 | 40100932 | ||
BC115500.1 | - | 1 | 303 | 40100933 |