Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EXOC7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013839.3 | NP_001013861.1 | 18 | 2013 | ||
NM_001145297.3 | NP_001138769.1 | 19 | 2166 | |||
NM_001145298.3 | NP_001138770.1 | 18 | 1989 | |||
NM_001145299.3 | NP_001138771.1 | 19 | 2082 | |||
NM_001282313.1 | NP_001269242.1 | 18 | 2042 | |||
NM_015219.4 | NP_056034.2 | 17 | 1920 | |||
XM_006721786.3 | XP_006721849.1 | 20 | 2120 | |||
XM_006721787.3 | XP_006721850.1 | 19 | 2486 | |||
XM_006721788.3 | XP_006721851.1 | 19 | 2027 | |||
XM_006721789.3 | XP_006721852.1 | 18 | 1958 | |||
XM_011524571.2 | XP_011522873.1 | 19 | 2397 | |||
XM_017024397.1 | XP_016879886.1 | 15 | 1522 | |||
GenBank mRNA | AB028990.1 | - | 18 | 1936 | ||
AK022552.1 | - | 9 | 704 | |||
AK023832.1 | - | 17 | 1828 | |||
AK098284.1 | - | 9 | 2185 | |||
AK128278.1 | - | 4 | 278 | |||
AK295871.1 | - | 18 | 1994 | |||
AL834324.1 | - | 18 | 1984 | |||
AY869728.1 | - | 15 | 1716 | |||
BC011045.1 | - | 18 | 1947 | 4215228 | ||
BC018466.1 | - | 19 | 2074 | 3867492 | ||
BX647763.1 | - | 16 | 1681 | |||
BX648435.1 | - | 2 | 569 | |||
FJ457119.1 | - | 18 | 1894 | |||
FJ457120.1 | - | 19 | 1987 |