Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dclk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001111051.1 | NP_001104521.1 | 13 | 4957 | ||
NM_001111052.1 | NP_001104522.1 | 14 | 5031 | |||
NM_001195538.1 | NP_001182467.1 | 17 | 5818 | |||
NM_001195539.1 | NP_001182468.1 | 13 | 5005 | |||
NM_019978.3 | NP_064362.1 | 17 | 5866 | |||
GenBank mRNA | AK034632.1 | - | 1 | 2832 |