Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNMBP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318326.1 | NP_001305255.1 | 13 | 3407 | ||
NM_001318327.1 | NP_001305256.1 | 13 | 2158 | |||
NM_015221.3 | NP_056036.1 | 16 | 4404 | |||
XM_006717735.2 | XP_006717798.1 | 15 | 4270 | |||
XM_011539559.2 | XP_011537861.1 | 17 | 4475 | |||
GenBank mRNA | AB023227.1 | - | 13 | 3494 | ||
AK128576.1 | - | 13 | 3406 | |||
AK303479.1 | - | 8 | 1505 | |||
AL833283.1 | - | 16 | 4370 | |||
AY196211.1 | - | 15 | 4278 | |||
BC041628.1 | - | 13 | 2147 | 4329464 |