Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Inadl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005784.1 | NP_001005784.1 | 10 | 1322 | ||
NM_172696.2 | NP_766284.2 | 10 | 1322 | |||
GenBank mRNA | AK050545.1 | - | 10 | 1294 | ||
AK136048.1 | - | 4 | 505 | |||
AK147866.1 | - | 10 | 1271 | |||
BC050846.1 | - | 10 | 1256 | |||
BC141404.1 | - | 10 | 1298 | 9056017 |