Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CALM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001305624.1 | NP_001292553.1 | 5 | 419 | ||
NM_001305625.1 | NP_001292554.1 | 4 | 278 | |||
NM_001305626.1 | NP_001292555.1 | 3 | 3633 | |||
NM_001743.5 | NP_001734.1 | 4 | 373 | |||
GenBank mRNA | AK055130.1 | - | 3 | 2090 | ||
AK315694.1 | - | 4 | 261 | |||
BC003354.1 | - | 4 | 249 | 2899899 | ||
BC006464.1 | - | 4 | 251 | 3542044 | ||
BC008437.1 | - | 4 | 267 | 4092978 | ||
BC017385.1 | - | 4 | 265 | 3681165 | ||
BC018677.1 | - | 4 | 266 | 4656551 | ||
BC026065.1 | - | 4 | 265 | 3683323 | ||
BG563041.1 | - | 5 | 313 | |||
BP370358.1 | - | 4 | 353 | |||
BT009916.1 | - | 3 | 193 | |||
CR541990.1 | - | 3 | 193 | |||
CR542021.1 | - | 3 | 193 | |||
D45887.1 | - | 4 | 261 | |||
HY022313.1 | - | 4 | 373 | |||
HY108355.1 | - | 4 | 279 |