Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmsb15l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207267.4 | NP_997150.1 | 4 | 401 | ||
GenBank mRNA | AK029707.1 | - | 4 | 378 | ||
BB623661.1 | - | 4 | 397 | |||
BC096054.1 | - | 4 | 334 | 3373135 | ||
BE625563.1 | - | 4 | 326 |
Gene Symbol: Tmsb15b2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080967.4 | NP_001074436.1 | 3 | 261 | ||
GenBank mRNA | CN836907.1 | - | 247 | |||
CN842750.1 | - | 3 | 264 |