Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gm20816 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001160144.1 | NP_001153616.1 | 2 | 218 | ||
GenBank mRNA | BC100413.1 | - | 2 | 218 | 30919580 |
Gene Symbol: Gm20747 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025241.3 | NP_001020412.1 | 2 | 117 | ||
GenBank mRNA | BC089498.1 | - | 2 | 117 | 6744023 |
Gene Symbol: Gm20815 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017394.2 | NP_001017394.2 | 218 |
Gene Symbol: Gm20823 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001160143.1 | NP_001153615.1 | 2 | 218 | ||
GenBank mRNA | BC089467.1 | - | 2 | 218 | 6772630 |
Gene Symbol: Gm20852 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001160129.1 | NP_001153601.1 | 2 | 148 | ||
GenBank mRNA | BC087891.1 | - | 2 | 148 | 6774426 |
Gene Symbol: Gm20867 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001160142.1 | NP_001153614.1 | 2 | 218 | ||
GenBank mRNA | BC089466.1 | - | 2 | 218 | 6742357 |
Gene Symbol: Gm20868 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK006494.1 | - | 2 | 203 |
Gene Symbol: Gm20877 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199332.1 | NP_001186261.1 | 2 | 211 |
Gene Symbol: Gm21943 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017393.3 | NP_001017393.1 | 2 | 196 | ||
GenBank mRNA | BC089465.1 | - | 2 | 196 | 6707203 |
Gene Symbol: Ssty2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023546.3 | NP_076035.3 | 2 | 205 | ||
GenBank mRNA | L04852.1 | - | 2 | 205 |