Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013602.3 | NP_038630.1 | 1 | 188 | ||
GenBank mRNA | AK007983.1 | - | 1 | 37 | ||
AK011040.1 | - | 1 | 37 | |||
AK018727.1 | - | 1 | 51 | |||
AK154954.1 | - | 1 | 37 | |||
BC027262.1 | - | 1 | 66 | 3483861 | ||
BC036990.1 | - | 33 | 4990006 | |||
BC109369.1 | - | 547 |