Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gm13139 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083918.1 | NP_001077387.1 | 7 | 846 | ||
GenBank mRNA | BC024323.1 | - | 7 | 846 | 4242170 |
Gene Symbol: Gm13251 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001085522.2 | NP_001078991.1 | 4 | 574 |
Gene Symbol: Znf41-ps | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC096621.1 | - | 629 | 30464103 |