Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nptxr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030689.4 | NP_109614.2 | 5 | 3925 | ||
GenBank mRNA | AK147546.1 | - | 5 | 3983 | ||
AK147605.1 | - | 5 | 3934 | |||
BC024758.1 | - | 1 | 1211 | |||
BC026413.1 | - | 1 | 822 | |||
BC048914.1 | - | 1 | 885 | |||
BC058962.1 | - | 5 | 3924 | 6826075 | ||
BC059870.1 | - | 1 | 1221 | |||
BC098199.1 | - | 5 | 3924 | 5705707 |
Gene Symbol: Npcd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013360.2 | NP_001013378.2 | 8 | 3558 | ||
NM_001013362.2 | NP_001013380.2 | 9 | 3849 | |||
GenBank mRNA | BC060093.1 | - | 1 | 1362 |