Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TSPYL4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021648.4 | NP_067680.3 | 1 | 975 | ||
GenBank mRNA | AB018264.1 | - | 1 | 944 | ||
AK091559.1 | - | 3 | 728 | |||
AK302542.1 | - | 1 | 978 | |||
BC009116.1 | - | 1 | 349 | |||
BC040453.1 | - | 1 | 919 |