Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nptxr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030689.4 | NP_109614.2 | 5 | 1424 | ||
GenBank mRNA | AF316612.1 | - | 5 | 1406 | ||
AK006611.1 | - | 3 | 385 | |||
AK147546.1 | - | 5 | 1481 | |||
AK147605.1 | - | 5 | 1432 | |||
AY394487.1 | - | 6 | 1132 | |||
BC058962.1 | - | 5 | 1423 | 6826075 | ||
BC098199.1 | - | 5 | 1423 | 5705707 | ||
BC141096.1 | - | 5 | 1403 | 9055708 |
Gene Symbol: Npcd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013360.2 | NP_001013378.2 | 8 | 1057 | ||
NM_001013362.2 | NP_001013380.2 | 9 | 1348 | |||
GenBank mRNA | AY394488.1 | - | 8 | 962 | ||
AY394489.1 | - | 9 | 1253 | |||
AY395907.1 | - | 11 | 1862 | |||
CD578976.1 | - | 4 | 663 |