Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM208A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001112736.1 | NP_001106207.1 | 23 | 4520 | ||
NM_015224.3 | NP_056039.2 | 16 | 3748 | |||
XM_005264999.1 | XP_005265056.1 | 23 | 4655 | |||
XM_006713077.2 | XP_006713140.1 | 23 | 4532 | |||
XM_006713078.2 | XP_006713141.1 | 17 | 3509 | |||
XM_011533552.2 | XP_011531854.1 | 22 | 4472 | |||
XM_011533553.2 | XP_011531855.1 | 22 | 4076 | |||
XM_017006030.1 | XP_016861519.1 | 23 | 4307 | |||
XM_017006031.1 | XP_016861520.1 | 22 | 4472 | |||
XM_017006032.1 | XP_016861521.1 | 22 | 3953 | |||
GenBank mRNA | AB029028.1 | - | 6 | 1601 | ||
AF180425.2 | - | 16 | 4032 | |||
AF273053.1 | - | 9 | 2672 | |||
AK307599.1 | - | 15 | 3210 | |||
BC070096.1 | - | 22 | 4336 | |||
BC129987.1 | - | 15 | 3296 | 40129237 | ||
BX648955.1 | - | 22 | 4002 |