Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gm10375 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098269.2 | NP_001091739.1 | 4 | 428 | ||
GenBank mRNA | BC089536.1 | - | 4 | 428 | 6742411 |
Gene Symbol: 1700001F09Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027940.2 | NP_082216.1 | 4 | 465 | ||
GenBank mRNA | AK005587.1 | - | 4 | 466 | ||
BC061060.1 | - | 4 | 447 | 6770724 |
Gene Symbol: BC061237 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198677.1 | NP_941079.1 | 4 | 480 | ||
GenBank mRNA | BC061237.1 | - | 4 | 480 | 6705671 |
Gene Symbol: Gm10377 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001244671.1 | NP_001231600.1 | 4 | 428 | ||
GenBank mRNA | BC049548.1 | - | 4 | 428 |