Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: H2afx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010436.2 | NP_034566.1 | 1 | 321 | ||
GenBank mRNA | AK008124.1 | - | 1 | 341 | ||
AK088040.1 | - | 1 | 344 | |||
BC005468.1 | - | 1 | 315 | 3490058 | ||
BC010336.1 | - | 1 | 321 | 3156946 | ||
X58069.1 | - | 1 | 325 |
Gene Symbol: H2afj | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177688.4 | NP_808356.1 | 1 | 366 | ||
GenBank mRNA | AK053755.1 | - | 1 | 291 | ||
AV498351.1 | - | 1 | 365 | |||
BC024397.1 | - | 1 | 326 | 5055276 | ||
BC099606.1 | - | 1 | 348 | 1383389 |
Gene Symbol: Hist1h2aa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175658.2 | NP_783589.1 | 1 | 274 | ||
GenBank mRNA | BC107354.1 | - | 1 | 274 | 40055855 | |
BC107355.1 | - | 1 | 274 | 40055860 |
Gene Symbol: Hist1h2ab | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175660.3 | NP_783591.2 | 1 | 319 | ||
GenBank mRNA | BC117110.1 | - | 1 | 318 | 40126429 |
Gene Symbol: Hist1h2ac | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178189.5 | NP_835496.1 | 1 | 316 | ||
GenBank mRNA | AK033518.1 | - | 318 |
Gene Symbol: Hist1h2ad | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178188.4 | NP_835495.1 | 1 | 364 | ||
GenBank mRNA | BC119295.1 | - | 1 | 364 | 8734044 | |
BC119297.1 | - | 1 | 364 | 8734046 |
Gene Symbol: Hist1h2ae | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178187.4 | NP_835494.1 | 1 | 386 | ||
GenBank mRNA | AK028026.1 | - | 1 | 387 | ||
BC058544.1 | - | 1 | 305 | 5711765 | ||
BC076498.1 | - | 1 | 304 | 5713252 |
Gene Symbol: Hist1h2af | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175661.2 | NP_783592.1 | 1 | 274 | ||
GenBank mRNA | BC125011.1 | - | 1 | 274 | 40051842 |
Gene Symbol: Hist1h2ag | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178186.3 | NP_835493.1 | 1 | 296 | ||
GenBank mRNA | BC065803.1 | - | 1 | 296 | 1263745 |
Gene Symbol: Hist1h2ah | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175659.2 | NP_783590.1 | 1 | 365 | ||
GenBank mRNA | BC125248.1 | - | 1 | 273 | ||
BC134370.1 | - | 1 | 274 | 40100857 | ||
BC139479.1 | - | 1 | 365 | 8862501 | ||
BC146008.1 | - | 1 | 365 | 40131311 |
Gene Symbol: Hist1h2ai | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178182.2 | NP_835489.1 | 1 | 284 | ||
GenBank mRNA | BC099406.1 | - | 284 | 30789778 | ||
BC116373.2 | - | 1 | 274 | 40100893 | ||
BC127164.1 | - | 1 | 274 | 40100895 |
Gene Symbol: Hist1h2ak | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178183.2 | NP_835490.1 | 1 | 359 | ||
GenBank mRNA | BC119335.1 | - | 1 | 359 | 8734084 |
Gene Symbol: Hist1h2al | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK077055.1 | - | 132 |
Gene Symbol: Hist1h2an | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178184.2 | NP_835491.1 | 1 | 274 |
Gene Symbol: Hist1h2ao | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177544.2 | NP_001171015.1 | 1 | 374 |
Gene Symbol: Hist1h2ap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178185.2 | NP_835492.1 | 1 | 274 | ||
GenBank mRNA | AK145443.1 | - | 1 | 307 | ||
AK146846.1 | - | 1 | 305 | |||
BC062251.1 | - | 1 | 286 | 1224905 | ||
BC090402.1 | - | 1 | 286 | 5150365 | ||
BC110626.1 | - | 1 | 274 | 40051841 | ||
BC133661.1 | - | 1 | 274 | 40051853 | ||
BC147161.1 | - | 1 | 374 | 9056189 | ||
BC147162.1 | - | 1 | 374 | 9056203 |
Gene Symbol: Hist2h2aa1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013549.2 | NP_038577.1 | 1 | 358 | ||
GenBank mRNA | AK002725.1 | - | 1 | 318 | ||
AK006728.1 | - | 1 | 317 | |||
BC010564.1 | - | 1 | 301 | 3582122 | ||
BC053966.1 | - | 1 | 310 | |||
BC062255.1 | - | 1 | 301 | 1448126 | ||
BC064002.1 | - | 300 | ||||
BC089519.1 | - | 1 | 358 | 6771160 | ||
BC147778.1 | - | 1 | 355 | 9087498 | ||
BC147781.1 | - | 1 | 355 | 9087501 | ||
Z30940.1 | - | 1 | 296 |
Gene Symbol: Hist2h2aa2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178212.3 | NP_835584.1 | 1 | 398 |
Gene Symbol: Hist2h2ac | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175662.2 | NP_783593.1 | 1 | 310 | ||
GenBank mRNA | BC117094.1 | - | 1 | 310 | 40126413 | |
BC117096.1 | - | 1 | 310 | 40126415 |
Gene Symbol: Hist3h2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178218.4 | NP_835736.1 | 1 | 327 | ||
GenBank mRNA | AK051448.1 | - | 328 | |||
AK077568.1 | - | 1 | 328 | |||
AK082083.1 | - | 331 | ||||
AK087537.1 | - | 329 | ||||
BC058119.1 | - | 300 | 6819179 | |||
BC063781.1 | - | 294 | 6493838 |