Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Defa-rs7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AJ564870.1 | - | 100 | |||
AJ564871.1 | - | 100 | ||||
AJ564872.1 | - | 100 | ||||
AJ564873.1 | - | 100 | ||||
AJ564875.1 | - | 100 | ||||
AJ564876.1 | - | 100 | ||||
AJ564877.1 | - | 100 |
Gene Symbol: Defa-rs2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AJ564864.1 | - | 100 | |||
AJ564866.1 | - | 100 | ||||
AJ564868.1 | - | 100 |
Gene Symbol: Defa-rs4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AJ564862.1 | - | 100 |