Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rps23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024175.3 | NP_077137.1 | 4 | 438 | ||
GenBank mRNA | AK002573.1 | - | 4 | 396 | ||
AK010594.1 | - | 4 | 396 | |||
AK010608.1 | - | 4 | 394 | |||
AK010667.1 | - | 4 | 408 | |||
AK012287.1 | - | 4 | 396 | |||
AK012438.1 | - | 4 | 390 | |||
AK012574.1 | - | 4 | 396 | |||
AK013720.1 | - | 4 | 397 | |||
AK050580.1 | - | 4 | 394 | |||
AK088085.1 | - | 4 | 396 | |||
AK166994.1 | - | 4 | 430 | |||
AK168587.1 | - | 4 | 390 | |||
BC002145.1 | - | 4 | 385 | 3484753 | ||
BC054435.1 | - | 3 | 375 | 5373967 | ||
BC078418.1 | - | 3 | 375 | 5329878 | ||
BC100603.1 | - | 4 | 397 | 6760902 | ||
BQ746768.1 | - | 4 | 376 |