Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hist1h4n | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175657.2 | NP_783588.1 | 1 | 181 | ||
GenBank mRNA | BC115448.2 | - | 1 | 181 | 40099764 | |
BC152397.1 | - | 1 | 181 | 40099768 |
Gene Symbol: Hist1h4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178192.2 | NP_835499.1 | 1 | 273 | ||
GenBank mRNA | AK016310.1 | - | 1 | 255 | ||
BC119241.1 | - | 1 | 273 | 8733990 | ||
BC119243.1 | - | 1 | 273 | 8733992 |
Gene Symbol: Hist1h4b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178193.2 | NP_835500.1 | 1 | 265 | ||
GenBank mRNA | BC132186.1 | - | 1 | 265 | 40130463 | |
BC132212.1 | - | 1 | 265 | 40130489 |
Gene Symbol: Hist1h4c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178208.2 | NP_835515.1 | 1 | 190 | ||
GenBank mRNA | BC115450.1 | - | 1 | 181 | 40099776 | |
BC115451.1 | - | 1 | 174 | |||
BC139809.1 | - | 1 | 181 | 40099779 |
Gene Symbol: Hist1h4d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175654.2 | NP_783585.1 | 1 | 181 | ||
GenBank mRNA | BC119611.2 | - | 1 | 180 |
Gene Symbol: Hist1h4f | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175655.2 | NP_783586.1 | 1 | 181 | ||
GenBank mRNA | BC115446.1 | - | 1 | 174 |
Gene Symbol: Hist1h4h | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153173.4 | NP_694813.1 | 1 | 252 | ||
GenBank mRNA | AA028777.1 | - | 1 | 250 | ||
AK007642.1 | - | 1 | 248 | |||
BC028550.1 | - | 1 | 193 | 1398106 | ||
BC058529.1 | - | 1 | 189 | 6817839 | ||
BC087952.1 | - | 1 | 241 | 6787571 |
Gene Symbol: Hist1h4i | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175656.3 | NP_783587.1 | 1 | 220 | ||
GenBank mRNA | AK011560.1 | - | 1 | 221 | ||
BC019757.1 | - | 1 | 210 | |||
BC052219.1 | - | 1 | 193 | |||
BC092144.1 | - | 1 | 210 | 30612926 | ||
BC125598.1 | - | 1 | 204 | 40130113 | ||
BC125600.1 | - | 1 | 204 | 40130115 |
Gene Symbol: Hist1h4j | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178210.1 | NP_835582.1 | 1 | 181 | ||
GenBank mRNA | AK010085.1 | - | 1 | 217 |
Gene Symbol: Hist1h4k | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178211.2 | NP_835583.1 | 1 | 229 | ||
GenBank mRNA | AK139521.1 | - | 1 | 216 | ||
BC096767.1 | - | 196 | ||||
BC117010.1 | - | 1 | 229 | 40126329 | ||
BC117012.1 | - | 1 | 229 | 40126331 | ||
BC119612.1 | - | 1 | 181 | 40061668 |
Gene Symbol: Hist1h4m | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195421.1 | NP_001182350.1 | 1 | 219 | ||
GenBank mRNA | BC139425.1 | - | 243 | 8862447 | ||
BC139437.1 | - | 243 | 8862459 |
Gene Symbol: Hist2h4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033596.3 | NP_291074.1 | 1 | 194 | ||
GenBank mRNA | BC051566.1 | - | 1 | 202 | ||
BC057955.1 | - | 188 | 5359141 | |||
BX520915.1 | - | 1 | 194 |