Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bglap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037939.2 | NP_001033028.1 | 3 | 532 | ||
NM_007541.3 | NP_031567.1 | 4 | 326 | |||
GenBank mRNA | CJ326308.1 | - | 4 | 327 | ||
CJ326768.1 | - | 1 | 330 | |||
U11542.1 | - | 4 | 293 |