Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLHL18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025010.4 | NP_079286.2 | 10 | 1731 | ||
XM_005265001.3 | XP_005265058.2 | 10 | 1661 | |||
XM_005265002.4 | XP_005265059.2 | 10 | 1646 | |||
XM_005265003.3 | XP_005265060.2 | 10 | 1646 | |||
XM_006713079.1 | XP_006713142.1 | 10 | 1617 | |||
XM_017006033.1 | XP_016861522.1 | 9 | 1524 | |||
XM_017006034.1 | XP_016861523.1 | 9 | 1509 | |||
XM_017006035.1 | XP_016861524.1 | 9 | 1494 | |||
GenBank mRNA | AB018338.1 | - | 8 | 1279 | ||
AB062478.1 | - | 10 | 1903 | |||
AK291477.1 | - | 10 | 1625 | |||
AK295292.1 | - | 9 | 1493 | |||
BC016388.2 | - | 6 | 901 | |||
BC032620.1 | - | 10 | 1543 | 5578173 | ||
BX537953.1 | - | 10 | 1622 |