Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mup9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281979.1 | NP_001268908.1 | 2 | 214 | ||
GenBank mRNA | BI219655.1 | - | 2 | 195 |
Gene Symbol: LOC100048884 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199333.1 | NP_001186262.1 | 2 | 220 | ||
GenBank mRNA | BC055353.1 | - | 195 |
Gene Symbol: Mup1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163010.1 | NP_001156482.1 | 2 | 259 | ||
NM_001163011.1 | NP_001156483.1 | 2 | 259 | |||
NM_031188.2 | NP_112465.2 | 2 | 259 | |||
GenBank mRNA | BC012221.1 | - | 2 | 205 | 4195753 | |
BC013649.1 | - | 193 | 4195401 | |||
BC091744.1 | - | 196 | 5102271 | |||
BC091775.1 | - | 2 | 206 | 5137341 | ||
BC092254.1 | - | 193 | 5100458 | |||
BC093515.1 | - | 193 | ||||
BC099597.1 | - | 196 | 947916 | |||
BC106092.1 | - | 2 | 196 | 5098340 | ||
M16355.1 | - | 2 | 259 | |||
M27609.1 | - | 61 | ||||
M28649.1 | - | 206 | ||||
X03524.1 | - | 61 | ||||
X04115.1 | - | 17 |
Gene Symbol: Mup10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001122647.1 | NP_001116119.1 | 2 | 215 | ||
GenBank mRNA | AJ309921.1 | - | 148 | |||
CB948918.1 | - | 124 | ||||
EU882231.1 | - | 2 | 148 |
Gene Symbol: Mup11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164526.1 | NP_001157998.1 | 246 | |||
GenBank mRNA | AK013972.1 | - | 2 | 223 |
Gene Symbol: Mup12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199995.1 | NP_001186924.1 | 2 | 214 |
Gene Symbol: Mup14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199999.1 | NP_001186928.1 | 2 | 257 |
Gene Symbol: Mup15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001200004.1 | NP_001186933.1 | 2 | 220 |
Gene Symbol: Mup16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199936.1 | NP_001186865.1 | 2 | 214 |
Gene Symbol: Mup19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135127.2 | NP_001128599.1 | 2 | 214 | ||
GenBank mRNA | AK013259.1 | - | 2 | 214 | ||
EU882232.1 | - | 2 | 142 |
Gene Symbol: Mup2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001045550.2 | NP_001039015.1 | 2 | 218 | ||
NM_008647.4 | NP_032673.3 | 2 | 218 | |||
GenBank mRNA | BC012259.1 | - | 207 | 4196403 | ||
BC059097.1 | - | 193 | 4159984 | |||
CB593503.1 | - | 216 | ||||
M16356.1 | - | 206 |
Gene Symbol: Mup8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134676.1 | NP_001128148.1 | 2 | 278 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BC094452.1 | - | 194 | Chr. - - [-] on Build |