Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gm15114 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001082966.1 | NP_001076435.1 | 7 | 1908 |
Gene Symbol: Gm15080 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001198988.1 | NP_001185917.1 | 7 | 1963 |
Gene Symbol: Gm15085 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001122734.1 | NP_001116206.1 | 1962 |
Gene Symbol: Gm15093 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099920.1 | NP_001093390.1 | 1956 |
Gene Symbol: Ott | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011022.1 | NP_035152.1 | 1962 | |||
GenBank mRNA | X96606.1 | - | 1962 |