Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLUH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015229.3 | NP_056044.3 | 15 | 2563 | ||
XM_011523768.2 | XP_011522070.1 | 14 | 2494 | |||
GenBank mRNA | AB014564.1 | - | 11 | 1952 | ||
AK023003.1 | - | 7 | 1413 | |||
AK125717.1 | - | 15 | 2583 | |||
BC063786.1 | - | 9 | 1645 | |||
CR627449.1 | - | 13 | 2301 |