Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcng2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107372.1 | NP_001100842.1 | 3 | 1602 | ||
XM_006254947.2 | XP_006255009.1 | 3 | 2976 | |||
XM_006254948.2 | XP_006255010.1 | 3 | 2146 | |||
XM_006254950.2 | XP_006255012.1 | 3 | 1497 | |||
XM_008772136.1 | XP_008770358.1 | 3 | 2113 | |||
XM_008772137.1 | XP_008770359.1 | 3 | 1541 | |||
GenBank mRNA | AJ011020.1 | - | 2 | 1468 |