Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc35a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127642.1 | NP_001121114.1 | 3 | 384 | ||
XM_006256693.2 | XP_006256755.1 | 3 | 666 | |||
GenBank mRNA | BC166811.1 | - | 3 | 384 | 7453742 |