Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tceb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270561.1 | NP_001257490.1 | 3 | 314 | ||
NM_001270562.1 | NP_001257491.1 | 3 | 212 | |||
NM_001270563.1 | NP_001257492.1 | 3 | 172 | |||
NM_022593.4 | NP_072115.1 | 3 | 208 | |||
XM_006237724.2 | XP_006237786.1 | 3 | 209 | |||
GenBank mRNA | BC060566.1 | - | 3 | 183 | 6918038 | |
CK355540.1 | - | 3 | 208 | |||
FM093546.1 | - | 3 | 174 | |||
FM101371.1 | - | 3 | 162 | |||
FM110992.1 | - | 3 | 178 | |||
FQ213660.1 | - | 3 | 222 | |||
L29259.1 | - | 2 | 125 |