Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rap2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053741.1 | NP_446193.1 | 1 | 87 | ||
GenBank mRNA | AY037826.1 | - | 1 | 87 |