Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Actr1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039028.2 | NP_001034117.2 | 3 | 278 | ||
GenBank mRNA | BC166803.1 | - | 3 | 245 | 7938869 | |
EV779654.1 | - | 3 | 306 | |||
FQ228979.1 | - | 3 | 246 |