Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rps19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037346.2 | NP_001032423.1 | 6 | 486 | ||
XM_006228411.1 | XP_006228473.1 | 6 | 453 | |||
GenBank mRNA | BC087641.1 | - | 6 | 458 | 7308408 | |
CB720565.1 | - | 6 | 486 | |||
FQ221309.1 | - | 6 | 468 | |||
FQ221510.1 | - | 6 | 469 | |||
FQ224282.1 | - | 6 | 469 | |||
FQ229780.1 | - | 6 | 469 | |||
X51707.1 | - | 6 | 468 |
Gene Symbol: LOC100910336 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003750647.3 | XP_003750695.1 | 6 | 541 | ||
XM_008766901.1 | XP_008765123.1 | 6 | 453 |
Gene Symbol: Rps19l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008765600.1 | XP_008763822.1 | 2 | 442 |