Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prss1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012635.1 | NP_036767.1 | 5 | 742 | ||
GenBank mRNA | FQ211333.1 | - | 5 | 745 | ||
FQ226369.1 | - | 5 | 743 | |||
FQ227841.1 | - | 5 | 743 | |||
FQ233648.1 | - | 5 | 743 | |||
FQ235029.1 | - | 5 | 743 | |||
V01273.1 | - | 5 | 742 |
Gene Symbol: LOC103690254 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008762925.1 | XP_008761147.1 | 5 | 730 |
Gene Symbol: Try4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108626.1 | NP_001102096.1 | 5 | 747 | ||
GenBank mRNA | FQ231537.1 | - | 5 | 744 |