Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Npb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153293.1 | NP_695205.1 | 1 | 213 | ||
GenBank mRNA | AB085944.1 | - | 1 | 213 |