Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spetex-2G | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009971.1 | NP_001009971.1 | 4 | 547 | ||
GenBank mRNA | AB180082.1 | - | 4 | 547 |
Gene Symbol: Spetex-2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009976.1 | NP_001009976.1 | 4 | 592 | ||
GenBank mRNA | AB180076.1 | - | 4 | 592 |
Gene Symbol: Spetex-2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011697.1 | NP_001011697.1 | 4 | 547 | ||
GenBank mRNA | AB180077.1 | - | 4 | 547 |
Gene Symbol: Spetex-2C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011698.1 | NP_001011698.1 | 4 | 547 | ||
GenBank mRNA | AB180078.1 | - | 4 | 547 |
Gene Symbol: Spetex-2E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011702.1 | NP_001011702.1 | 547 | |||
GenBank mRNA | AB180080.1 | - | 547 |
Gene Symbol: Spetex-2F | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009968.1 | NP_001009968.1 | 4 | 547 | ||
GenBank mRNA | AB180081.1 | - | 4 | 547 |
Gene Symbol: Spetex-2H | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009969.1 | NP_001009969.1 | 4 | 553 | ||
GenBank mRNA | AB180083.1 | - | 4 | 553 |