Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC688924 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109519.1 | NP_001102989.1 | 3 | 156 | ||
XM_008759189.1 | XP_008757411.1 | 3 | 198 | |||
XM_008759190.1 | XP_008757412.1 | 3 | 186 | |||
XM_008759191.1 | XP_008757413.1 | 4 | 226 | |||
XM_008759192.1 | XP_008757414.1 | 4 | 342 | |||
XM_008759193.1 | XP_008757415.1 | 3 | 169 | |||
XM_008759194.1 | XP_008757416.1 | 3 | 261 | |||
XM_008759195.1 | XP_008757417.1 | 3 | 145 |