Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem107 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109648.1 | NP_001103118.1 | 3 | 359 | ||
XM_006246808.2 | XP_006246870.1 | 3 | 604 | |||
XM_006246811.2 | XP_006246873.1 | 3 | 578 | |||
XM_006246812.2 | XP_006246874.1 | 3 | 545 | |||
XM_008767875.1 | XP_008766097.1 | 3 | 583 |